Learn More
Restek™ Stainless Steel Fittings
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Ideal for connecting stainless steel tubing in UHPLC systems
$23.75 - $49.50
Specifications
Material | Stainless Steel |
---|---|
For Use With (Application) | 1/16 in. Tubing |
Port Style | 10-32 |
Quantity | 5/Pk. |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
06-611-712
![]() |
Restek™
27731 |
Rheodyne-Style Nuts (Short) |
Pack of 5 for $41.00
|
|
|||||
06-611-713
![]() |
Restek™
27732 |
Rheodyne-Style Nuts (Medium) |
Pack of 5 for $42.50
|
|
|||||
06-611-714
![]() |
Restek™
27733 |
Rheodyne-Style Nuts (Long) |
Pack of 5 for $49.50
|
|
|||||
06-611-716
![]() |
Restek™
27735 |
Rheodyne-Style Ferrules (Cone-Style) |
Pack of 5 for $30.50
|
|
|||||
06-611-715
![]() |
Restek™
27734 |
Rheodyne-Style Ferrules (Stepped-Style) |
Pack of 5 for $36.25
|
|
|||||
06-611-708
![]() |
Restek™
27727 |
Valco-Style Nuts (Short) |
Pack of 5 for $23.75
|
|
|||||
06-611-709
![]() |
Restek™
27728 |
Valco-Style Nuts (Medium) |
Pack of 5 for $30.50
|
|
|||||
06-611-710
![]() |
Restek™
27729 |
Valco-Style Nuts (Long) |
Pack of 5 for $35.00
|
|
|||||
06-611-711
![]() |
Restek™
27730 |
Valco Ferrule |
Pack of 5 for $36.25
|
|
|||||
Description
- For 1/16 in. tubing on all 10-32 ports
- 1/4 in. head on all nuts
- Ferrules not included, must be purchased separately
Specifications
Stainless Steel | |
1/16 in. Tubing | |
10-32 | |
5/Pk. |