Learn More
MilliporeSigma™ Supelco™ SSI™ High Pressure Precolumn Filter
In-line installation. The 316 stainless steel filter disc (0.5 μm pores) is easily replaced without removing the column end fitting. For 1/16 in. tubing. Maximum operating pressure: 15,000 psi (1,054 kg/cm2).
Supplier: MilliporeSigma™ Supelco™ 59269
Specifications
Fitting: thread 10-32 Valco compatible |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SSI is a trademark of Scientific Systems, Inc.