Learn More
TARGETMOL CHEMICALS INC SR-3029 10MG
Supplier: TARGETMOL CHEMICALS INC T3972
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR-3029 is a potent and highly specific CK1(delta)/CK1(epsilon) inhibitor. purity: 100%