Learn More
Electron Microscopy Sciences Sputtering Target Gold-Palladium (60:40), 39 mm
Supplier: Electron Microscopy Sciences 91009AP
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
EMS precious metal sputtering targets are made from high purity metals. The targets are for use on most sputtering units and includes systems manufacturers Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and others. Our targets come in a standard thickness of 0.1 mm (100um). Targets are available mounted or unmounted. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you.
EMS strives to maintain accurate pricing. However, due to fluctuations in precious metal prices, pricing on products containing precious metals is not guaranteed. We will contact you if there is a discrepancy.
Diameter: 39 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.