Learn More
Sklar Instruments Sponge/Utility Bowl, Sklar, Plastic. This bowls measures 6-3/8 in. x 3 in. and has a capacity of 1-1/4 quarts. EA/1.
SDP
Supplier:  Sklar Instruments 101502
Encompass_Preferred
Sponge/Utility Bowl, Sklar, Plastic. Sklar Sponge Bowls are most commonly utilized as a recepticle for contaminated sponges to be placed throughout a surgical procedure. They may; however, also be used as a container for holding sponges or sponge sticks that have been soaked in a sterile solution to be used during a procedure. This bowls measures 6-3/8 in. x 3 in. and has a capacity of 1-1/4 quarts.
Catalog No. 50-143-7011
Explore available promotions