Learn More
Quantimetrix Spinalscopics, Spinal Fluid Cell Count Control, Level 2
Supplier: Quantimetrix 146231
•Liquid, human protein matrix with stabilized human cells•24-month refrigerated shelf life from date of manufacture•6-month open vial stability when stored at 2ºC–8ºC•1-month open vial stability at room temperatureMonitor the performance of red and white blood cell count methods in cerebrospinal fluid samples performed manually using a hemocytometer. Easy-to-use dropper dispenser.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.