Learn More
Quantimetrix Dropper Spinal Fluid Control, Level 1
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Quantimetrix 145131
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
•Easy-to-use dropper bottles•Liquid, human protein matrix•2-year refrigerated shelf life from date of manufacture•90-day open vial stability when stored at 2ºC–8ºCMonitor the performance of spinal fluid chemistry assay methods with Dropper Spinal Fluid Control. Drop the control directly into the instrument sample cup with an easy-to-use, dropper bottle for simple dispensing and maximum use. Dropper Spinal Fluid Control is designed for use with most major chemistry analyzers and nephelometers including: Siemens Dimension, Beckman Coulter AU series, Roche Cobas, Abbott Architect and Pyrogallol Red microprotein methods. Dropper Spinal Fluid Control Analytes: Albumin, Chloride, Glucose, IgG, Lactic Acid, LDH, Sodium, Total Protein
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.