Learn More
Terra™ Spider Low Men's CT FP Black Boot
Engineered to stabilize lateral movement, the Spider features a low-profile direct-injected PU midsole and outsole with supportive sidewall chassis. For all-day comfort, the Spider comes equipped with a premium PU FOOTBED™ and CLEANFEET™ odor-control treated liner.
Supplier: Terra™ R8115B 060 M
Description
- Premium water resistant suede and full-grain leather
- CLEANFEET™ treated moisture wicking performance lining
- Reflective heel accents
- TERRA FIRMA-FLEX™ Metal Free technology in toe and plate
- Direct injected low profile PU midsole and outsole, with sidewall chassis
- Removable CLEANFEET™ treated PU FOOTBED™
Specifications
Black | |
6 | |
No |
Nubuck | |
Men's | |
Work Boot |