Learn More
Perkin Elmer US LLC SPB Digital Controller
Supplier: Perkin Elmer US LLC N9300808
Includes features such as: Displays block or sample temperature Displays actual time remaining in sequence Select from 10 languages Utilizes Safety Messages to prevent failure Timer with shut-off option Temperature set-point to 0.1 Alarm for end of segment Calibration routine for DigiPROBE