Learn More
Biolegend Spark YGTM 581 anti-human CD20
Supplier: Biolegend 329238
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Spark YG™ 581 anti-human CD200 (OX2) [OX-104]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests