Learn More
Bel-Art™ SP Scienceware™ OdoClave™ Deodorant Autoclave Pads
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Neutralize autoclave odors with a pleasant fragrance
Supplier: Bel-Art™ H131990000/EMD
Description
- Heat-release deodorant works automatically at autoclave temperatures
- Open pouch and place in bag prior to autoclaving
- Three-year shelf life or longer if stored in a sealed container
- Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads
Specifications
Triple-strength Clove | |
Autoclavable | |
Yellow |