Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SouthernBiotech™ Genetically Distinct Collagens: Bovine Type VI
Highly purified fibrous protein for a variety of applications
$219.00 - $437.00
Specifications
For Use With (Application) | Cell Culture Coating Material, Antigen for Antibody Production, Collagen Gel Formation, Collagen Standard |
---|---|
Purification Method | Controlled and limited pepsin digestion, followed by selective salt precipitation |
Source | Bovine |
Product Type | Collagen, Bovine Type VI |
Content And Storage | Store at 2°C to 8°C or in small aliquots at -20°C |
Catalog Number | Mfr. No. | Quantity | Source | Purification Method | Purity or Quality Grade | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Source | Purification Method | Purity or Quality Grade | Price | Quantity & Availability | ||
OB1300-02S
|
SouthernBiotech™
130002S |
0.25 mg | Bovine | Controlled and limited pepsin digestion, followed by selective salt precipitation | >90% by SDS-PAGE |
Each for $437.00
|
|
||
OB1300-02
|
SouthernBiotech™
130002 |
0.1 mg | Bovine | Controlled and limited pepsin digestion, followed by selective salt precipitation | >90% by SDS-PAGE |
Each for $219.00
|
|
||
Description
Bovine origin, Type VI.
- Can be used as a collagen standard, antigen for antibody production, and coating material in cell culture studies, and in the formation of collagen gels
- Prepared via controlled and limited pepsin digestion; greater than 90% pure
- Supplied as a 0.5mg/mL solution in 500mmol/L acetic acid or lyophilized
Specifications
Cell Culture Coating Material, Antigen for Antibody Production, Collagen Gel Formation, Collagen Standard | |
Bovine | |
Store at 2°C to 8°C or in small aliquots at -20°C |
Controlled and limited pepsin digestion, followed by selective salt precipitation | |
Collagen, Bovine Type VI | |
>90% by SDS-PAGE |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title