Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SouthernBiotech™ Genetically Distinct Collagens: Bovine Type II
Highly purified fibrous protein for a variety of applications
Supplier: SouthernBiotech™ 122002
Description
Bovine origin, Type II.
- Can be used as a collagen standard, antigen for antibody production, and coating material in cell culture studies, and in the formation of collagen gels
- Prepared via controlled and limited pepsin digestion; greater than 90% pure
- Supplied as a 0.5mg/mL solution in 500mmol/L acetic acid or lyophilized
Specifications
Cell Culture Coating Material, Antigen for Antibody Production, Collagen Gel Formation, Collagen Standard | |
0.25 mg | |
Collagen, Bovine Type II | |
Lyophilized |
Controlled and limited pepsin digestion, followed by selective salt precipitation | |
Bovine | |
Store at 2°C to 8°C or in small aliquots at -20°C | |
>90% by SDS-PAGE |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction