Learn More
Cole-Parmer Extech SL130W Sound Level Meter with Led Alert and 110V Adapter
Supplier: Cole-Parmer 5050989
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These sound level meters not only meet ANSI Type 2 and ENIEC60651 specifications, they put the data to work for you. The large LCD display has high contrast digits and a bargraph and has LED indicator that can be viewed from 100 feet whether placed on a desk or mounted on a wall.