Learn More
Cole-Parmer Extech 407744 Sound Level Calibrator, 94db
Supplier: Cole-Parmer 5050986
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Use with sound meters featuring a 0.5" to 1.0" (12.7 to 25.4 mm) microphone.