Learn More
Hanna Pharmaceutical Supply Co., Inc. Sodasorb LF CO2 Absorbent
Supplier: Hanna Pharmaceutical Supply Co., Inc. 008865
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
1.2kg Refill Bag. Permanent color change when absorbent is exhausted Specifically formulated to prevent the degradation of anesthetic gases that can lead to problematic byproducts Exhibits high contrast color change, should dessication occur Long lasting formulation More efficient use of anesthetic gases Uniform pellet design results in less dust, better absorption
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.