Learn More
Cole-Parmer SNOL 13/1100 LHM01 Muffle Furnace, 13 L, Upwards Opening Door, 120V
Supplier: Cole-Parmer 3385918
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Using high quality thermoinsulation, ceramic hearth plates, and embedded heating elements on models up to 1100°C (exposed on ceramic tubes on models up to 1300°C) these furnaces provide excellent temperature stability and uniformity. Equipped with high-precision digital microprocessor controllers fitted with self-tuning and manual PID settings, they are an optimal fit for scientific, educational, government, industrial, and material testing labs.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.