Learn More
Thermo Scientific™ SLIPFREE Ferrules
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
Specifications
For Use With (Equipment) | SLIPFREE Connectors for HPLC Columns |
---|---|
Type | Sealing Ferrule |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
03-170-505
|
Thermo Scientific™
36024 |
Vespel |
Each of 1 for $30.10
|
N/A | |||||
Description
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
- Vespel ferrules replace the standard Vespel ferrules supplied with SLIPFREE column connectors
- Kel-F and PEEK are offered for applications in which Vespel is not suitable
Specifications
SLIPFREE Connectors for HPLC Columns | |
Sealing Ferrule |