Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sirtinol, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Cell-permeable, selective sirtuin deacetylase inhibitor (IC50 values are 38, 68 and 131 μM at SIRT2, Sir2p and SIRT1 respectively) that has no effect on HDAC1 activity. Significantly decreases growth and viability of PCa and HEK293T cells in vitro.
Specifications
Quantity | 50 mg |
Formula Weight | 394.47 |
Percent Purity | >98% |
Chemical Name or Material | Sirtinol |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction