Learn More
ILEX LIFE SCIENCES LLC Biocolor Sircol Collagen Acid-Salt Wash Reagent Concentrate, Component, 3 x 20 ml
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: ILEX LIFE SCIENCES LLC SWASH
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Biocolor Sircol™ Soluble Collagen Assay is a quantitative assay for measurement of both acid-soluble and pepsin-soluble collagens. The colorimetric assay is a dye-binding method using Sircol™ Dye Reagent, which contains Sirius Red dye in picric acid. This kit is suitable for all soluble mammalian collagens, Types I to V. The assay can assess the rate of newly synthesized collagen. New collagen is produced during periods of rapid growth and development. New collagen is also generated during inflammation, wound healing and tumor development. The Sircol™ Assay can monitor collagen produced in situ, or during in-vitro cell culture. It can also measure in-vitro extracellular matrix (ECM) formation. Sensitivity: 1.0 µg. Assay Run Time: 1.5 hours. NOTE: This assay is not suitable for covalent cross-linked collagen.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.