Learn More
Electron Microscopy Sciences Veco Grid L2 x 1.5, Nickel with 2000 x 1500 µm Slot
Supplier: Electron Microscopy Sciences 2015NI
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Slot: 2000 x 1500 µm
Material: Nickel