Learn More
Electron Microscopy Sciences Gilder Single Slot Oval Grid GS2/3x1, Gold, 606 x 1000 µm Slot
Supplier: Electron Microscopy Sciences G60610AU
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Gold
Slot: 606 x 1000 µm