Learn More
Electron Microscopy Sciences Silicone Carbide Powder, 10 µm particles 1 LB
Supplier: Electron Microscopy Sciences 5026010
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Dark gray 98% SiC. Hardness, Moh's scale 9.0; Knoop Scale 2500. Crystallography: hexagonal–alpha and beta phase.
Size: 10 µm