Learn More
SiliCycle™ SiliaPlate™ TLC Plates, Aluminum-backed, Silica
Aluminum-backed TLC plates provide quick, inexpensive handling for versatile and classical separations.
$159.70 - $369.00
Specifications
Backing Material Aluminum
Material Aluminum
Particle Size 10 to 12 μm
Type Aluminum-backed TLC Plates
For Use With (Application) TLC Analysis
View More Specs
Products 8
Catalog Number Mfr. No. Thickness (Metric) Fluorescent Indicator Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Thickness (Metric) Fluorescent Indicator Dimensions (L x W) Price Quantity & Availability  
01-184-083
View Documents
SiliCycle™
TLAR10011B124
200 μm F254 2.5 x 7.5 cm
Pack of 200 for $283.00
Add to Cart
01-184-082
View Documents
SiliCycle™
TLAR10011B005
150 μm F254 4 x 8 cm
Pack of 50 for $195.10
Add to Cart
01-184-087
View Documents
SiliCycle™
TLAR10011B515
200 μm F254 5 x 10 cm
Pack of 50 for $159.70
Add to Cart
01-184-086
View Documents
SiliCycle™
TLAR10011B415
200 μm F254 5 x 20 cm
Pack of 50 for $226.50
Add to Cart
01-184-088
View Documents
SiliCycle™
TLAR10011B712
200 μm F254 10 x 20 cm
Pack of 20 for $232.00
Add to Cart
01-184-084
View Documents
SiliCycle™
TLAR10011B323
200 μm F254 20 x 20 cm
Pack of 25 for $269.50
Add to Cart
01-184-085
View Documents
SiliCycle™
TLAR10011B323N
200 μm No 20 x 20 cm
Pack of 25 for $369.00
Add to Cart
01-184-089
View Documents
SiliCycle™
TLAR10014BK1112
200 μm F254 5 x 7.5 cm Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
SilicaPlate™ Aluminum-backed TLC plates provide an irregular particle shape and are low-weight.
  • Thin plates
  • High heating stability
  • Low fragility
  • Possible to cut with scissors
  • Can be stored in notebook
Specifications
Specifications
Aluminum
10 to 12 μm
TLC Analysis
Bare Silica (Silicon Dioxyde, SiO2)
Aluminum
Aluminum-backed TLC Plates
SiliaPlate