Learn More
Sigma Diagnostics™ Sigmacote™
Reacts with surface silanol groups on glass to produce a neutral, hydrophobic, microscopically thin film
Supplier: Sigma Diagnostics™ SL2100ML/DUP
Description
- Clear, colorless solution of a chlorinated organopolysiloxane in heptane
Specifications
Colorless | |
Siliconizing Reagent for Glass and Other Surfaces | |
100 mL |
2°C to 8°C | |
Used to treat GC injection glass inserts. | |
Solution |