Learn More
Epredia™ Shandon™ CytoRich™ Red Collection Fluid
Allow for more accurate evaluation of cytological features with Epredia™ Shandon™ CytoRich™ Red Collection Fluid, which lyses RBCs and solubilizes proteins.
Supplier:  Epredia™ B9990800
 View more versions of this product
Catalog No. B9990800
Add to Cart
Description
Description
  • Pre-filled containers available
  • Ready-to-use, alcohol-based fluid
  • Compatible with immunohistochemistry
Specifications
Specifications
Collection Fluid
1 gal.
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov