Learn More
Epredia™ Cryochrome™ Embedding Resin
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
Supplier: Epredia™ 9990422
Description
- Color codes frozen sections for easy identification and tracking
- Four colors available: pink, blue, yellow, green
- Formulated with physical characteristics identical to Cryomatrix™ embedding resin
Specifications
Embedding Media | |
120mL |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov