Learn More
TARGETMOL CHEMICALS INC SGI-7079 10MG
Supplier: TARGETMOL CHEMICALS INC T6982
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SGI-7079 is a new selective Axl inhibitor. SGI-7079 can be a dose-dependent manner inhibited growth of tumors. It is also a potential therapeutic target for EGFR inhibitor resistance. purity: 99%