Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SGI 1027
DNA methyltransferase inhibitor
$286.52 - $1177.40
Specifications
Purity | >99% |
---|---|
Product Type | SGI 1027 |
Molecular Weight (g/mol) | 461.52 |
Inhibitors | SGI 1027 |
Description
DNA methyltransferase inhibitor. Inhibits the mammalian DNA methyltransferases DNMT3B, DNMT3A and DNMT1 (IC50 values are 7.5, 8 and 12.5 μM with Poly(dl-dC) as the substrate). Reactivates silenced tumor suppressor genes by reducing CpG island hypermethylation.Specifications
>99% | |
SGI 1027 | |
461.52 | |
SGI 1027 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title