Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Series 330 Face Shield Replacement Windows
Click to view available options
Hard cap adapter
Description
Uvex™ Bionic™ is a revolutionary face shield designed for rugged jobs. Extremely lightweight and highly adjustable for all-day comfort, it is perfect for workers exposed to impact, chemical splash, and airborne debris.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Gray |
Product Type | Hard Cap Adaptor |
For Use With (Equipment) | Mounts on hard cap |
Includes | Ratchet suspension |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction