Learn More
Thermo Scientific™ Sensititre™ Gram Negative RUO Susceptibility Testing Plate
NEW antimicrobial options including cefiderocol and imipenem/relebactam together with colistin and fosfomycin for difficult to treat Gram-negative infections in a high-quality, ready to use AST broth microdilution (BMD) plate format, meeting the latest CLSI breakpoints.
Supplier: Thermo Scientific™ YMDRGNX2F
Description
Expand your Gram-negative organism susceptibility testing options by accessing the latest antibiotics including cefiderocol and imipenem/relebactam. The Thermo Scientific™ Sensititre™ MDRGNX2F plate delivers results directly from broth microdilution (BMD), the gold standard1 phenotypic method for Antimicrobial Susceptibility Testing (AST). Open access to one of the most comprehensive range of second-line antimicrobials, together with colistin and fosfomycin on a single plate format, designed for research use only.
- Updated standard format including cefiderocol, imipenem/relebactam, colistin and fosfomycin
- Research Use Only (RUO)
- BMD provides accurate MIC results therefore negating the need for repeat testing
- Designed for use with CLSI breakpoints
- Sensititre MIC determination is achieved using direct measurement rather than relying on extrapolated data, thereby providing an accurate MIC result, not an estimate.
- All contained antibiotics also available in customizable plate options, along with access to over 300 antimicrobials, to enable meeting local formulary and Stewardship program requirements
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Various | |
Various | |
Multidrug-resistant, Gram-negative Organisms | |
10/Pk. |
Sensititre Gram Negative RUO AST Plate 10/Pk | |
AST Plate | |
Minimum Inhibitory Concentration (MIC) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.