Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ SecurAlert Color Card Clip-on Badge
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Product Type:
One-day expiration
One-week expiration
For Use With (Application):
Contractors
Temporary
Visitors
Description
- Low-cost clip-on alternative to self-adhesive badges
- Package contains 500 front and back parts
- Self-expiring cards prevent unauthorized use
Specifications
Width (English) | 3 in. |
For Use With (Application) | Temporary |
Product Type | One-week expiration |
Dimensions (L x W) | 3 in. x 3 in. |
Includes | Two parts per card |
Color | Green |
Length (English) | 3 in. |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction