Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
Supplier: Thermo Scientific™ SPD300DDA230
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
- For strong acids and bases used in combinatorial chemistry
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
UL Listed to US and Canadian requirements. CE marked | |
Aluminum | |
35°C to 85°C |
SpeedVac SPD300DDA Vacuum concentrator | |
SPD300 | |
230 V 50 Hz |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.