Learn More
Sartorius arium™ Pro ASTM TYPE 1 - Ultrapure Water System, Benchtop
![GREENER_CHOICE](/content/dam/fishersci/glyphs/greener_choice.png)
Produce ASTM type 1 water for critical analysis like HPLC, PCR, antibody production, cell culture media preparation or electrophoresis by adding a arium™ Pro ASTM TYPE 1 - Ultrapure Water System to your lab.
$9376.00 - $11673.00
Specifications
Certifications/Compliance | ASTM |
---|---|
Bacterial Content | <1 CFU/L |
Product Water Type | Type 1 |
Feedwater Temperature (English) | 35.6°F to 86°F |
Feedwater Temperature (Metric) | 2°C to 30°C |
Catalog Number | Mfr. No. | Type | Total Organic Carbon | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Total Organic Carbon | Price | Quantity & Availability | ||||
14-556-025
![]() |
Sartorius
H2OPROUVTTOC |
Arium Pro ASTM Type 1 Ultrapure UV Benchtop Water System | <2 product water, <50 feed water |
Each for $11,673.00
|
|
||||
14-556-212
|
Sartorius
H2OPRODIT |
Arium Pro ASTM Type 1 Ultrapure DI Benchtop Water System | <5 product water, <50 feed water |
Each for $9,376.00
|
|
||||
14-556-017
|
Sartorius
H2OBASICT/X |
Arium Pro ASTM Type 1 Ultrapure Basic Benchtop Water System | <5 product water, <50 feed water | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
The arium pro series consist of flexible systems with device configurations that can be specially custom-tailored to applications and feature an outstanding cost-benefit ratio. All systems meet and exceed ASTM Type 1 water quality and ensure the best, reproducible results. Per hour, an arium pro can deliver up to 120 l of ultrapure water with a conductivity of 0.055μS/cm (18.2 megaohm/cm at 25°C) and consistently high quality. When a Sartopore° 2 150 final filter is used, arium pro will deliver ultrapure water that is practically free of microorganisms.
- Multiple configurations offered based on application needs
- Bench integrated design
- Optimized components and cartridge materials with topdown-flow technology
- Optional, integrated UV lamp (185/254nm) for eliminating bacteria and organics; controlled by reliable TOC monitoring
- Display with touch-activated functions and intuitive menu navigation
- Integrated real time monitoring and data display: feed conductivity, total water consumption, pressure regulator
- PIN-protected access to basic settings /service
- Convenient SD card slot
- System can be chosen based on daily work
- Guarantees a long service life and optimal water purification
- Reliable water quality ensures reproducible results
- Easy operation; no user manual needed
- Optimal control of constant water flow and consistent water quality without extra costs
- Prevents user operating errors
- Supports reliable recording of important data
Specifications
ASTM | |
Type 1 | |
2°C to 30°C | |
Benchtop | |
4 to 10 | |
5°C to 45°C | |
2°C to 35°C | |
13.7 in. | |
19.37 in. | |
17.75 in. | |
59.5 to 63.9 lb. | |
100/240 V 50/60 Hz | |
50/60 Hz |
<1 CFU/L | |
35.6°F to 86°F | |
80% | |
<1/mL | |
Adsorption by spherical activated Carbon, deionization, optional final particle and sterile filtration | |
35.6°F to 95°F | |
<1 | |
35 cm | |
49.2 cm | |
45.1 cm | |
27 to 29 kg | |
100/240 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.