Learn More
Sino Biological SARS-CoV-2 Spike RBD (L452Q, F490S) Protein (His & AVI Tag), Biotinylated 20ug
Supplier: Sino Biological 40592V49H6B20
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
A DNA sequence encoding the SARS-CoV-2 Spike RBD (YP_009724390.1, with mutations L452Q, F490S) (Arg319-Phe541) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed. The mutations were identified in the SARS-CoV-2 variant (known as C.37) which emerged in the Peru.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.