Learn More
Scilogex LLC SAFEVAC ASPIRATION SYSTEM,110V
Supplier: Scilogex LLC 761000019999
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Integrated vacuum pump with quick & easy controlVacuum power can be set to different levels to achieve 1mL/S to 15mL/ S aspiration speedQuick connectors for easy installationFully autoclavable waste collection bottle, tubing and handleSensitive level sensor to prevent liquid overflow during operationSingle channel nozzle (supplied) & 8-channel nozzle accessoryLid, tubing and handle are easy for disassembling, cleaning and autoclaving
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.