Learn More
Glass Expansion Safety Gloves, Small
Supplier: Glass Expansion SAFEGLOVESML
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Safety Gloves, small. These safety gloves are provided with the D-Torch for use while handling the D-Torch. Always wear safety gloves and safety glasses when handling ICP torches because they contain glass. Other safety glove sizes available are medium (P/N SAFEGLOVE-med) and large (P/N SAFEGLOVE-lge). Safety glasses are also available (P/N SAFEGLASS). Safety first!Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.