Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aqua Phoenix™ Safe Racer 2.0 Deluxe Kit Components
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Replacement parts for Educational Kit
Description
Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.Specifications
Elastic cord | |
K to 5 | |
Elastic | |
1 mm |
Clear | |
Education Kit | |
7 m |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction