Learn More
Zymo Research Corporation R-Zymolyase (1000 Units Lyophilized) w/ Storage Buffer (500 µl)
SDP
Supplier:  Zymo Research Corporation E1006
Encompass_Preferred
This special formula of enzymes is specially manufactured for yeast genomic DNA preparations. Our liquid Zymolyase is prepared from Arthrobacter luteus. The primary yeast lytic activity is 1,3-glucan laminaripentaohydrolase. It hydrolyzes glucose polymers at the is -1,3-glucan linkages releasing laminaripentaose as the principal product. This formula may be used as Zymolyase as it is, but be cautious that it contains RNase A.
Catalog No. 50-444-505
Explore available promotions