Learn More
TARGETMOL CHEMICALS INC RUPATADINE FUMARATE 500MG
Supplier: TARGETMOL CHEMICALS INC T6242
Also available in 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rupatadine Fumarate (Rinialer) (UR-12592 Fumarate) is a potent dual PAF/H1 antagonist with Ki of 0.55/0.1 uM. purity: 99%