Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One Round Bottom Polystyrene Test Tubes
Clear, round bottom test tubes for sample transfer.
Supplier: Greiner Bio-One 151101
Description
Clear, round bottom test tubes for sample transfer.
Polystyrene tubes are ideally suited for optical measurements as a result of its high clarity.
- Nonsterile
- Available in different sizes
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Clear | |
65 mm | |
Sample transfer | |
16 mm |
6.5 mL | |
Polystyrene | |
Non-sterile | |
Test Tube |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction