Learn More
Medchemexpress LLC HY-18257 5mg Medchemexpress, Rolitetracycline CAS:751-97-3 Purity:>98%
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Medchemexpress LLC HY182575MG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Medchemexpress, HY-18257 5mg Rolitetracycline CAS:751-97-3 Rolitetracycline, a derivative of tetracycline, is a broad-spectrum antibiotic [1] [2] . Rolitetracyclin has a role as a protein synthesis inhibitor, an antiprotozoal drug and a prodrug [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.