Learn More
World Precision Instrument Rib Cutting Knife, 25 mm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument WP0089
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Rib Cutting Knife is designed for gross anatomy dissection and post-mortem exams. It has a thick cutting edge and a sturdy, balance handle for cutting through ribs. Made of standard grade stainless steel, it is easily sanitized with any disinfectant or autoclaved between uses.