Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris™ Rhein
FTO (mRNA N6-methyladenine demethylase) inhibitor; also inhibits ALKBH2 and ALKBH3
Specifications
Molecular Formula | C15H8O6 |
---|---|
Formula Weight | 284.22 |
Product Type | Inhibitor |
Purity | 98% |
Description
Rhein is a FTO (mRNA N6-methyladenine demethylase) inhibitor (IC50 = 1.2 - 21 μM; Ki = 1.8 μM). Competitively binds enzyme at active site preventing binding of N6-methyladenosine (m6A) substrate and resulting in accumulation of DNA damage in cells. Also inhibits ALKBH3 and ALKBH2 (IC50 values are 5.3 and 9.1 μM, respectively). Sensitizes glioblastoma cells to alkylating agents. Active in vivo.Specifications
C15H8O6 | |
284.22 | |
Inhibitor | |
98% |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title