Learn More
Medchemexpress LLC HY-111498 5mg Medchemexpress, RGX-104 CAS:610318-03-1 Purity:>98%
Supplier: Medchemexpress LLC HY1114985MG
Medchemexpress, HY-111498 5mg RGX-104 CAS:610318-03-1 RGX-104 is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.