Learn More
Restek™ Ultra HPLC Columns with C18 Column Packing, Trident Inlet Fitting
SDP
Similar in pore size to Pinnacle II™ columns, but with greater retention
Supplier:  Restek™ 9174562700
 View more versions of this product
Catalog No. 06-716-344
Add to Cart
Description
Description
  • High-purity silica contains only trace amounts of metal impurities, providing excellent selectivity without the need for mobile phase modifiers
  • With large surface area and high-density bondings
  • 2.5 to 7.5 pH range
  • Temperature limit: 80°C (176°F)
  • C18 columns are good for carbonyls, fatty acids, glycerides, anilines, fat-soluble vitamins, phthalates, barbiturates, steroids, PTH, amino acids, and other acids
Specifications
Specifications
Analytical Column
Trident™ inlet fitting
5 μm
100 Å
L1
Type B Silica
Reversed Phase
Reversed Phase
2.1 mm
150 mm
2.5 to 8
80°C
C18
20%
Yes
Product Suggestions
Product Suggestions