Learn More
Restek™ Stabilwax™ Capillary Columns
SDP
Long column lifetime, even when programming repeatedly up to 260°C (500°F)
Supplier:  Restek™ 106556850
 View more versions of this product
Catalog No. 06-709-118
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Fused silica
  • Polar phase; Crossbond™ polyethylene glycol
  • Rugged enough to withstand repeated water injections
  • Lowest-bleed PEG column on the market; long column lifetimes
  • Produces polar compound retention times that do not shift
  • Temperature range: 40°C to 260°C (104°F to 500°F)
  • Equivalent to USP G14, G15, G16, G20 and G39 phases
Applications:
Used for a wide range of compounds and matrices including: FAMEs, flavor compounds, essential oils, solvents, aromatics (including xylene isomers), acrolein/acrylonitrile (EPA 603) and oxygenated compounds. Also used for purity testing of chemicals and analyzing impurities in water matrices and alcoholic beverages.
Specifications
Specifications
Stabilwax™
Carbowax Polymer
Essential Oils Analysis, Flavor Compounds Analysis, Essential Oils, Solvents Analysis, Xylene Isomers Analysis
40°C to 240/260°C
30 m
Polar
Cage
0.53 mm
G14, G15, G16, G20, G39
1 μm
1 μm
Stainless Steel
Fused Silica
Product Suggestions
Product Suggestions