Learn More
Enzo Life Sciences Renin (human), (recombinant) (active) (10µg)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Enzo Life Sciences ENZPRT1930010
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Alternative name: Angiotensinogenase, Angiotensin forming enzyme, Ren. Concentration: 0.1 mg/ml when reconstituted as suggested. Purity: ≥90%. Formulation: Lyophilized. In 25mM MES buffer containing 150mM NaCl. Source: Produced in HEK cells. Mature Renin (aa 67-406) is fused at the N-terminus to a FLAG-tag. Active form, does not contain prorenin peptide (aa 1-66). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.