Learn More
Thermo Scientific™ Remel™ Color Slide™ RF Rapid Latex Test
Latex agglutination slide test for the qualitative and semi-quantitative determination of circulating rheumatoid factor (RF) in human serum
Supplier: Thermo Scientific™ R2471336
Includes: Positive and negative controls, latex reagent, sample diluent, pipets, and disposable test cards
Description
- CLIA Complexity: Non-Waived
- CPT Codes: 86431
- Utilizes mono-dispersed latex reagent
- Semi-quantitative titers can be performed directly on the reaction card for efficiency and accuracy
- No sample dilution is required for qualitative procedure
- Positive control is assayed to assist in regulatory compliance to prove proficiency of titering techniques
- Results in uniformly smooth negatives
- Test results within one minute
- For in vitro diagnostic use
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)