Learn More
ILEX LIFE SCIENCES LLC REC HUMAN ALBUMIN POWDER 1G
SDP
Supplier:  ILEX LIFE SCIENCES LLC PB020001G
Encompass_Preferred
Provis Biolabs Recombinant Human Albumin (rHA), Lyophilized Powder, GMP Grade (1 g). DESCRIPTION: Recombinant Human Albumin (rHA), GMP Grade, expressed in Pichia pastoris, is a 66 kDa synthetic alternative to human serum albumin (HSA). The rHA is a highly purified product that is free of animal or human components. It is developed as an alternative to HSA, to which it is structurally equivalent. The rHA offers a reliable and sustainable alternative to traditional HSA sources. Purity levels ≥ 99% confirmed by orthogonal quality control tests. Raw materials used are animal component free, TSE/BSE-free certified, and comply with IP/USP/EP. APPLICATIONS: Cell and tissue culture media supplement, used as serum substitute in serum-free xeno-free cell culture, medium component for cryopreservation and stem cell culture, stabilizer protein for pharmaceutical and biotech products, reduces unspecific binding in molecular biology assays.
Catalog No. 50-273-3643
Explore available promotions